accessibility menu, dialog, popup

UserName

Viewing profile as user:
Br

CAS RN 2051-98-1

IUPAC Name: 5-bromo-1,2-dihydroacenaphthylene
Synonyms: 5-bromo-1,2-dihydroacenaphthylene
Molecular Weight (g/mol): 233.11
Molecular Formula: C12H9Br
InChi Key: QALKJGMGKYKMKE-UHFFFAOYSA-N
SMILES: BrC1=C2C=CC=C3CCC(C=C1)=C23
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)