accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20511-12-0

IUPAC Name: 5-iodopyridin-2-amine
Synonyms: 5-iodopyridin-2-amine
Molecular Weight (g/mol): 220.01
Molecular Formula: C5H5IN2
InChi Key: IVILGUFRMDBUEQ-UHFFFAOYSA-N
SMILES: NC1=CC=C(I)C=N1
Molecular Weight (g/mol) 
  • (1)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (1)
Quantity 
  • (3)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (6)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)