accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2052-07-5

Br

CAS RN 2052-07-5

IUPAC Name: 2-bromo-1,1'-biphenyl
Synonyms: O-bromobiphenyl
Molecular Weight (g/mol): 233.11
Molecular Formula: C12H9Br
InChi Key: KTADSLDAUJLZGL-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1C1=CC=CC=C1
Boiling Point 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results