accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 205391-02-2

IUPAC Name: 3'-(acetyloxy)-5,6-diamino-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthen]-6'-yl acetate
Synonyms: 3'-(acetyloxy)-5,6-diamino-3-oxospiro[2-benzofuran-1,9'-xanthen]-6'-yl acetate
Molecular Weight (g/mol): 446.42
Molecular Formula: C24H18N2O7
InChi Key: PTSUYDXEEKDBQU-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC2=C(C=C1)C1(OC(=O)C3=CC(N)=C(N)C=C13)C1=C(O2)C=C(OC(C)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results