accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 205445-51-8

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-(3,4-difluorophenyl)propanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-(3,4-difluorophenyl)propanoic acid
Molecular Weight (g/mol): 301.29
Molecular Formula: C14H17F2NO4
InChi Key: CYAOPVHXASZUDE-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC1=CC(F)=C(F)C=C1)C(O)=O
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)