accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20548-54-3

IUPAC Name: calcium sulfanediide
Synonyms: calcium sulfide
Molecular Weight (g/mol): 72.14
Molecular Formula: CaS
InChi Key: AGVJBLHVMNHENQ-UHFFFAOYSA-N
SMILES: [S--].[Ca++]
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results