accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO(R)NNHOO

CAS RN 205526-34-7

IUPAC Name: (2R)-3-(4-cyanophenyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoate
Synonyms: (2R)-3-(4-cyanophenyl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoate
Molecular Weight (g/mol): 411.44
Molecular Formula: C25H19N2O4
InChi Key: JOPKKUTWCGYCDA-HSZRJFAPSA-M
SMILES: [O-]C(=O)[C@@H](CC1=CC=C(C=C1)C#N)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results