accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO(S)SNNHOO

CAS RN 205528-32-1

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(1,3-thiazol-4-yl)propanoate
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(1,3-thiazol-4-yl)propanoate
Molecular Weight (g/mol): 393.44
Molecular Formula: C21H17N2O4S
InChi Key: LSBAZFASKHLHKB-IBGZPJMESA-M
SMILES: [O-]C(=O)[C@H](CC1=CSC=N1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)