accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 205672-25-9

IUPAC Name: 5-bromo-2-chloropyrimidin-4-amine
Synonyms: 5-bromo-2-chloropyrimidin-4-amine
Molecular Weight (g/mol): 208.44
Molecular Formula: C4H3BrClN3
InChi Key: QOWALNIZDHZTSM-UHFFFAOYSA-N
SMILES: NC1=NC(Cl)=NC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results