accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 205676-17-1

IUPAC Name: (2Z)-2-(4-chlorophenyl)-3-hydroxyprop-2-enal
Synonyms: (2Z)-2-(4-chlorophenyl)-3-hydroxyprop-2-enal
Molecular Weight (g/mol): 182.6
Molecular Formula: C9H7ClO2
InChi Key: BPCQPZCKKNFWCF-VMPITWQZSA-N
SMILES: O\C=C(/C=O)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results