accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3O

CAS RN 20577-61-1

IUPAC Name: methyl 2,4-dioxopentanoate
Synonyms: methyl 2,4-dioxopentanoate
Molecular Weight (g/mol): 144.13
Molecular Formula: C6H8O4
InChi Key: OMHOEQINEXASKE-UHFFFAOYSA-N
SMILES: COC(=O)C(=O)CC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results