accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNOOBr

CAS RN 2058-72-2

IUPAC Name: 5-bromo-1-methyl-2,3-dihydro-1H-indole-2,3-dione
Synonyms: 5-bromo-1-methylindole-2,3-dione
Molecular Weight (g/mol): 240.06
Molecular Formula: C9H6BrNO2
InChi Key: GEEDYJPPYNIZLX-UHFFFAOYSA-N
SMILES: CN1C(=O)C(=O)C2=CC(Br)=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results