accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOSNH3C

CAS RN 20582-55-2

IUPAC Name: ethyl 4-methyl-1,3-thiazole-5-carboxylate
Synonyms: ethyl 4-methyl-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol): 171.21
Molecular Formula: C7H9NO2S
InChi Key: WISQBJLUORKXNY-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)N=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results