accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOHOCl(E/Z)

CAS RN 20595-47-5

IUPAC Name: 3-(2,5-dichlorophenyl)prop-2-enoic acid
Synonyms: 3-(2,5-dichlorophenyl)prop-2-enoic acid
Molecular Weight (g/mol): 217.05
Molecular Formula: C9H6Cl2O2
InChi Key: LYYBVUOVYNSRSE-UHFFFAOYSA-N
SMILES: OC(=O)C=CC1=C(Cl)C=CC(Cl)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results