accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20600-22-0

IUPAC Name: 1-bromo-4-(phenoxymethyl)benzene
Synonyms: 1-bromo-4-(phenoxymethyl)benzene
Molecular Weight (g/mol): 263.13
Molecular Formula: C13H11BrO
InChi Key: KZVLWVAXLYUCLW-UHFFFAOYSA-N
SMILES: BrC1=CC=C(COC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results