accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClHNOS

CAS RN 20600-44-6

IUPAC Name: 5-chloro-2,3-dihydro-1,3-benzothiazol-2-one
Synonyms: 5-chloro-3H-1,3-benzothiazol-2-one
Molecular Weight (g/mol): 185.63
Molecular Formula: C7H4ClNOS
InChi Key: NOVHYVKPKWACML-UHFFFAOYSA-N
SMILES: ClC1=CC=C2SC(=O)NC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results