accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20605-01-0

IUPAC Name: 1,3-diethyl 2,2-bis(hydroxymethyl)propanedioate
Synonyms: 1,3-diethyl 2,2-bis(hydroxymethyl)propanedioate
Molecular Weight (g/mol): 220.22
Molecular Formula: C9H16O6
InChi Key: WIOHBOKEUIHYIC-UHFFFAOYSA-N
SMILES: CCOC(=O)C(CO)(CO)C(=O)OCC
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (3)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (6)
  • (4)
  • (3)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)