accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2062-98-8

IUPAC Name: 2,3,3,3-tetrafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propanoyl fluoride
Synonyms: 2,3,3,3-tetrafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propanoyl fluoride
Molecular Weight (g/mol): 332.05
Molecular Formula: C6F12O2
InChi Key: BCLQALQSEBVVAD-UHFFFAOYNA-N
SMILES: FC(=O)C(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)