accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 206261-66-7

IUPAC Name: 2-[2-(dimethylamino)ethoxy]benzoic acid
Synonyms: 2-[2-(dimethylamino)ethoxy]benzoic acid
Molecular Weight (g/mol): 209.25
Molecular Formula: C11H15NO3
InChi Key: JZLYEZGHBQFRLU-UHFFFAOYSA-N
SMILES: CN(C)CCOC1=CC=CC=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)