accessibility menu, dialog, popup

UserName

Viewing profile as user:
BH3CCH3CH3CH3CH3H3CHO

CAS RN 20631-84-9

IUPAC Name: bis(2,4,6-trimethylphenyl)borinic acid
Synonyms: bis(2,4,6-trimethylphenyl)borinic acid
Molecular Weight (g/mol): 266.19
Molecular Formula: C18H23BO
InChi Key: OTYVTANMHIUXBQ-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(B(O)C2=C(C)C=C(C)C=C2C)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results