accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20634-92-8

IUPAC Name: ethyldipropylamine
Synonyms: ethyl dipropylamine
Molecular Weight (g/mol): 129.25
Molecular Formula: C8H19N
InChi Key: XWCCTMBMQUCLSI-UHFFFAOYSA-N
SMILES: CCCN(CC)CCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results