accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOO

CAS RN 2065-23-8

IUPAC Name: methyl 2-phenoxyacetate
Synonyms: methyl phenoxyacetate
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: BZCKRPHEZOHHBK-UHFFFAOYSA-N
SMILES: COC(=O)COC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results