accessibility menu, dialog, popup

UserName

Viewing profile as user:
IH3CP

CAS RN 2065-66-9

IUPAC Name: methyltriphenylphosphanium iodide
Synonyms: methyltriphenylphosphanium iodide
Molecular Weight (g/mol): 404.23
Molecular Formula: C19H18IP
InChi Key: JNMIXMFEVJHFNY-UHFFFAOYSA-M
SMILES: [I-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (8)
Quantity 
  • (4)
  • (4)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (3)
Physical Form 
  • (3)
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (8)
  • (4)
  • (4)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)