accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20651-67-6

IUPAC Name: 1-butyl-4-iodobenzene
Synonyms: 1-butyl-4-iodobenzene
Molecular Weight (g/mol): 260.12
Molecular Formula: C10H13I
InChi Key: XILRUONFYBUYIE-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(I)C=C1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results