CAS RN 206551-41-9
IUPAC Name:
methyl 3-bromo-2-fluorobenzoate
Synonyms:
methyl 3-bromo-2-fluorobenzoate
Molecular Weight (g/mol):
233.04
Molecular Formula:
C8H6BrFO2
InChi Key:
ZWOFHFOFKBYRHV-UHFFFAOYSA-N
SMILES:
COC(=O)C1=C(F)C(Br)=CC=C1
1
–
2
of
2
results
1
Promotions Available
Methyl 3-bromo-2-fluorobenzoate, 98%, Thermo Scientific Chemicals