CAS RN 20661-21-6
IUPAC Name:
indiganetriol
Synonyms:
indium(III) hydroxide
Molecular Weight (g/mol):
168.86
Molecular Formula:
H6InO3
InChi Key:
ZMFWDTJZHRDHNW-UHFFFAOYSA-N
SMILES:
O.O.O.[In]
1
–
2
of
2
results
1
Indium(III) hydroxide, 99.8% (metals basis)
CAS: 20661-21-6 MDL Number: MFCD00011056
2
Indium(III) hydroxide, 99.998% (metals basis)
CAS: 20661-21-6 MDL Number: MFCD00011056