accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20662-14-0

IUPAC Name: scandium(3+) hexahydrate trichloride
Synonyms: scandium(3+) hexahydrate trichloride
Molecular Weight (g/mol): 259.4
Molecular Formula: Cl3H12O6Sc
InChi Key: FKZFOHABAHJDIK-UHFFFAOYSA-K
SMILES: O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Sc+3]
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (3)
  • (2)
Grade 
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (4)
  • (4)