accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20662-84-4

IUPAC Name: trimethyl-1,3-oxazole
Synonyms: trimethyl-1,3-oxazole
Molecular Weight (g/mol): 111.14
Molecular Formula: C6H9NO
InChi Key: ZRLDBDZSLLGDOX-UHFFFAOYSA-N
SMILES: CC1=NC(C)=C(C)O1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)