accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20666-12-0

IUPAC Name: sodium 8-amino-4-oxo-3,4-dihydrophthalazin-1-olate
Synonyms: sodium 8-amino-4-oxo-3H-phthalazin-1-olate
Molecular Weight (g/mol): 199.15
Molecular Formula: C8H6N3NaO2
InChi Key: GBOXDRMCEVPUSV-UHFFFAOYSA-M
SMILES: [Na+].NC1=C2C([O-])=NNC(=O)C2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results