accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20686-65-1

IUPAC Name: 2-methylnaphtho[2,1-d][1,3]oxazole
Synonyms: 2-methylnaphtho[2,1-d][1,3]oxazole
Molecular Weight (g/mol): 183.21
Molecular Formula: C12H9NO
InChi Key: JKYSRHYQAVELLH-UHFFFAOYSA-N
SMILES: CC1=NC2=C(O1)C1=CC=CC=C1C=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results