accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2069-71-8

IUPAC Name: N1,N1-dimethyl-4-nitrobenzene-1,3-diamine
Synonyms: N1,N1-dimethyl-4-nitrobenzene-1,3-diamine
Molecular Weight (g/mol): 181.2
Molecular Formula: C8H11N3O2
InChi Key: WJTOMXLUNDWLCY-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C(N)=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results