accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH3COOH3COOH3COCe

CAS RN 206996-60-3

IUPAC Name: cerium(3+) triacetate
Synonyms: cerium triacetate
Molecular Weight (g/mol): 317.25
Molecular Formula: C6H9CeO6
InChi Key: VGBWDOLBWVJTRZ-UHFFFAOYSA-K
SMILES: [Ce+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)