accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20712-16-7

IUPAC Name: 5-chloro-3-methylpyridin-2-amine
Synonyms: 5-chloro-3-methylpyridin-2-amine
Molecular Weight (g/mol): 142.59
Molecular Formula: C6H7ClN2
InChi Key: QTZBTBLHYPSFMG-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=CN=C1N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results