accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20726-63-0

IUPAC Name: acetic acid; diethylamine
Synonyms: acetic acid; diethylamine
Molecular Weight (g/mol): 133.19
Molecular Formula: C6H15NO2
InChi Key: UEHUZQKLOWYOMO-UHFFFAOYSA-N
SMILES: CC(O)=O.CCNCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results