accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 207290-72-0

IUPAC Name: 5-(4-hydroxypiperidin-1-yl)thiophene-2-carbaldehyde
Synonyms: 5-(4-hydroxypiperidin-1-yl)thiophene-2-carbaldehyde
Molecular Weight (g/mol): 211.28
Molecular Formula: C10H13NO2S
InChi Key: QGNLQHONMNFZJP-UHFFFAOYSA-N
SMILES: OC1CCN(CC1)C1=CC=C(S1)C=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results