accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOHOHOHOHH2ONa

CAS RN 207300-70-7

IUPAC Name: sodium 2,3,4,5-tetrahydroxy-6-oxohexanoate hydrate
Synonyms: sodium 2,3,4,5-tetrahydroxy-6-oxohexanoate hydrate
Molecular Weight (g/mol): 234.14
Molecular Formula: C6H11NaO8
InChi Key: ICQRTPLATMRWJI-UHFFFAOYNA-M
SMILES: O.[Na+].OC(C=O)C(O)C(O)C(O)C([O-])=O
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)