CAS RN 207300-83-2
IUPAC Name:
ethyl (2S)-2-(2-azaniumylacetamido)-3-(4-hydroxyphenyl)propanoate
Synonyms:
ethyl (2S)-2-(2-ammonioacetamido)-3-(4-hydroxyphenyl)propanoate
Molecular Weight (g/mol):
267.3
Molecular Formula:
C13H19N2O4
InChi Key:
ZWDVBDLGFUJMJV-NSHDSACASA-O
SMILES:
CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)C[NH3+]