accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20734-76-3

IUPAC Name: 2-amino-4-methoxyphenol
Synonyms: 2-amino-4-methoxyphenol
Molecular Weight (g/mol): 139.15
Molecular Formula: C7H9NO2
InChi Key: TUADYTFWZPZZTP-UHFFFAOYSA-N
SMILES: COC1=CC(N)=C(O)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)