accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 207386-92-3

IUPAC Name: 8-hydroxyquinoline-5-sulfonic acid
Synonyms: 8-hydroxyquinoline-5-sulfonic acid
Molecular Weight (g/mol): 225.22
Molecular Formula: C9H7NO4S
InChi Key: LGDFHDKSYGVKDC-UHFFFAOYSA-N
SMILES: OC1=C2N=CC=CC2=C(C=C1)S(O)(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)