accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20739-58-6

IUPAC Name: oct-2-yn-1-ol
Synonyms: 2-octyn-1-ol
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: TTWYFVOMGMBZCF-UHFFFAOYSA-N
SMILES: CCCCCC#CCO
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)