accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOHOOH(E/Z)

CAS RN 207461-99-2

IUPAC Name: 1-(1,2-diphenylethyl)piperidine; but-2-enedioic acid
Synonyms: diphenidine
Molecular Weight (g/mol): 381.47
Molecular Formula: C23H27NO4
InChi Key: AEHZEDLXDLPQQD-UHFFFAOYNA-N
SMILES: OC(=O)C=CC(O)=O.C(C(N1CCCCC1)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results