accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2075-45-8

IUPAC Name: 4-bromo-1H-pyrazole
Synonyms: 4-bromopyrazole
Molecular Weight (g/mol): 146.98
Molecular Formula: C3H3BrN2
InChi Key: WVGCPEDBFHEHEZ-UHFFFAOYSA-N
SMILES: BrC1=CNN=C1
Boiling Point 
  • (1)
  • (3)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (8)
Percent Purity 
  • (3)
  • (2)
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
Physical Form 
  • (5)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (6)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (8)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (5)
  • (3)