accessibility menu, dialog, popup

UserName

Viewing profile as user:
OPOOONaNa

CAS RN 207569-06-0

IUPAC Name: disodium naphthalen-1-yl phosphate
Synonyms: disodium naphthalen-1-yl phosphate
Molecular Weight (g/mol): 268.12
Molecular Formula: C10H7Na2O4P
InChi Key: QYURIFWAOPAPAJ-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)