accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 207596-32-5

IUPAC Name: palladium(2+) dinitrate
Synonyms: palladium(II) nitrate
Molecular Weight (g/mol): 230.43
Molecular Formula: N2O6Pd
InChi Key: GPNDARIEYHPYAY-UHFFFAOYSA-N
SMILES: [Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (6)
  • (6)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)