CAS RN 20765-98-4
Rhodium(III) chloride hydrate, 38% Rh
CAS: 20765-98-4 Molecular Formula: Cl3Rh Molecular Weight (g/mol): 209.26 MDL Number: MFCD00149839 InChI Key: SONJTKJMTWTJCT-UHFFFAOYSA-K IUPAC Name: rhodium(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[Rh+3]
Rhodium(III) chloride hydrate, Rh 38.0-45.5%
CAS: 20765-98-4 Molecular Formula: Cl3Rh Molecular Weight (g/mol): 209.26 MDL Number: MFCD00149839 InChI Key: SONJTKJMTWTJCT-UHFFFAOYSA-K Synonym: Rhodium trichloride hydrate IUPAC Name: rhodium(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[Rh+3]
Rhodium(III) chloride hydrate, 99.95%, (trace metal basis)
CAS: 20765-98-4 Molecular Formula: Cl3Rh Molecular Weight (g/mol): 209.26 MDL Number: MFCD00149839 InChI Key: SONJTKJMTWTJCT-UHFFFAOYSA-K IUPAC Name: rhodium(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[Rh+3]
Rhodium AA Standard, 1000 ppm Rh in 3% HCl, Ricca Chemical
CAS: 20765-98-4 Molecular Formula: Cl3Rh Molecular Weight (g/mol): 209.26 MDL Number: MFCD00149839 InChI Key: SONJTKJMTWTJCT-UHFFFAOYSA-K IUPAC Name: rhodium(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[Rh+3]
Rhodium ICP Standard, 10,000 ppm Rh in 20% HCl, Ricca Chemical
CAS: 20765-98-4 Molecular Formula: Cl3Rh Molecular Weight (g/mol): 209.26 MDL Number: MFCD00149839 InChI Key: SONJTKJMTWTJCT-UHFFFAOYSA-K IUPAC Name: rhodium(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[Rh+3]
Rhodium ICP Standard, 1000 ppm Rh in 3% HCl, Ricca Chemical
CAS: 20765-98-4 Molecular Formula: Cl3Rh Molecular Weight (g/mol): 209.26 MDL Number: MFCD00149839 InChI Key: SONJTKJMTWTJCT-UHFFFAOYSA-K IUPAC Name: rhodium(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[Rh+3]
Rhodium ICP-MS Standard, 1000 ppm Rh in 3% HCl, Ricca Chemical
CAS: 20765-98-4 Molecular Formula: Cl3Rh Molecular Weight (g/mol): 209.26 MDL Number: MFCD00149839 InChI Key: SONJTKJMTWTJCT-UHFFFAOYSA-K IUPAC Name: rhodium(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[Rh+3]
ICP-MS Internal Standard, 100 ppm Rh in 3% HCl, Ricca Chemical
CAS: 20765-98-4 Molecular Formula: Cl3Rh Molecular Weight (g/mol): 209.26 MDL Number: MFCD00149839 InChI Key: SONJTKJMTWTJCT-UHFFFAOYSA-K IUPAC Name: rhodium(3+) trichloride SMILES: [Cl-].[Cl-].[Cl-].[Rh+3]
Sigma Aldrich Rhodium(III) chloride hydrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Rhodium(III) Chloride Hydrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Rhodium(III) Chloride Hydrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.