accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20766-85-2

IUPAC Name: 6-(prop-2-enamido)hexanoic acid
Synonyms: 6-(prop-2-enamido)hexanoic acid
Molecular Weight (g/mol): 185.22
Molecular Formula: C9H15NO3
InChi Key: SAQWCPXBLNGTCC-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCNC(=O)C=C
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results