accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20776-67-4

IUPAC Name: 2-amino-5-chloro-3-methylbenzoic acid
Synonyms: 2-amino-5-chloro-3-methylbenzoic acid
Molecular Weight (g/mol): 185.61
Molecular Formula: C8H8ClNO2
InChi Key: KOPXCQUAFDWYOE-UHFFFAOYSA-N
SMILES: CC1=C(N)C(=CC(Cl)=C1)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results