accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20779-13-9

IUPAC Name: 1,5-dimethyl naphthalene-1,5-disulfonate
Synonyms: 1,5-dimethyl naphthalene-1,5-disulfonate
Molecular Weight (g/mol): 316.34
Molecular Formula: C12H12O6S2
InChi Key: HCXPJMZQMWIBMO-UHFFFAOYSA-N
SMILES: COS(=O)(=O)C1=CC=CC2=C(C=CC=C12)S(=O)(=O)OC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results