accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 207801-20-5

IUPAC Name: sodium 4-methylbenzene-1-sulfinate
Synonyms: sodium p-toluenesulfinate
Molecular Weight (g/mol): 178.18
Molecular Formula: C7H7NaO2S
InChi Key: KFZUDNZQQCWGKF-UHFFFAOYSA-M
SMILES: [Na+].CC1=CC=C(C=C1)S([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)