accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 207801-29-4

IUPAC Name: yttrium(3+) tris(2,4-dioxopentan-3-ide)
Synonyms: yttrium(3+) triacetylacetonate
Molecular Weight (g/mol): 386.23
Molecular Formula: C15H21O6Y
InChi Key: CKIVBSKECCTIAG-UHFFFAOYSA-N
SMILES: [Y+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results